siRNA Details
| Gene Symbol: IGSF22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173588.3 | NP_775859.3 | 13 | 1979 | ||
| XM_011520030.2 | XP_011518332.1 | 11 | 4883 | |||
| XM_011520031.2 | XP_011518333.1 | 11 | 4883 | |||
| XM_017017607.1 | XP_016873096.1 | 12 | 4774 | |||
| XM_017017608.1 | XP_016873097.1 | 12 | 2797 | |||
| GenBank mRNA | AK095113.1 | - | 13 | 1979 | ||
| BC115396.1 | - | 13 | 1930 | |||
| BC115397.1 | - | 13 | 1930 | 40068579 | ||

IGSF22
Genomic Map