siRNA Details
| Gene Symbol: B4GALNT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173593.3 | NP_775864.3 | 17, 18 | 2599 | ||
| GenBank mRNA | AB089940.1 | - | 17, 18 | 2599 | ||
| AK097681.1 | - | 4, 5 | 984 | |||
| AK131277.1 | - | 4, 5 | 1990 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

B4GALNT3| Gene Symbol: B4GALNT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173593.3 | NP_775864.3 | 17, 18 | 2599 | ||
| GenBank mRNA | AB089940.1 | - | 17, 18 | 2599 | ||
| AK097681.1 | - | 4, 5 | 984 | |||
| AK131277.1 | - | 4, 5 | 1990 | |||