siRNA Details
| Gene Symbol: ADGRD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198827.3 | NP_942122.2 | 3, 4 | 752 | ||
| XM_005253566.2 | XP_005253623.1 | 3, 4 | 752 | |||
| GenBank mRNA | AY532280.1 | - | 3, 4 | 366 | ||
| BC112307.1 | - | 3, 4 | 223 | 8327775 | ||
| BC112309.1 | - | 3, 4 | 223 | 8327777 | ||
| EU523118.1 | - | 3, 4 | 267 | |||
More Information
Set Membership: |
|

ADGRD1
Genomic Map