siRNA Details
| Gene Symbol: CLEC9A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207345.3 | NP_997228.1 | 7 | 1054 | ||
| GenBank mRNA | AY358265.1 | - | 7 | 1054 | ||
| EU339276.1 | - | 4 | 441 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CLEC9A| Gene Symbol: CLEC9A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207345.3 | NP_997228.1 | 7 | 1054 | ||
| GenBank mRNA | AY358265.1 | - | 7 | 1054 | ||
| EU339276.1 | - | 4 | 441 | |||