siRNA Details
| Gene Symbol: CLEC9A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207345.3 | NP_997228.1 | 9 | 1253 | ||
| GenBank mRNA | AY358265.1 | - | 9 | 1253 | ||
| EU339276.1 | - | 6 | 640 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CLEC9A| Gene Symbol: CLEC9A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207345.3 | NP_997228.1 | 9 | 1253 | ||
| GenBank mRNA | AY358265.1 | - | 9 | 1253 | ||
| EU339276.1 | - | 6 | 640 | |||