siRNA Details
| Gene Symbol: GAS2L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303130.1 | NP_001290059.1 | 9 | 1332 | ||
| NM_001303131.1 | NP_001290060.1 | 10 | 1819 | |||
| NM_174942.2 | NP_777602.1 | 10 | 1453 | |||
| XM_005268804.3 | XP_005268861.1 | 11 | 1522 | |||
| XM_011538219.2 | XP_011536521.1 | 10 | 1378 | |||
| XM_011538220.2 | XP_011536522.1 | 9 | 2713 | |||
| XM_011538221.2 | XP_011536523.1 | 9 | 1563 | |||
| XM_017019206.1 | XP_016874695.1 | 11 | 1596 | |||
| GenBank mRNA | AK095594.1 | - | 10 | 1819 | ||
| AK315189.1 | - | 9 | 1310 | |||
| BC043366.1 | - | 10 | 1403 | 5104644 | ||
| BX649059.1 | - | 1 | 268 | |||

GAS2L3
Genomic Map