siRNA Details
| Gene Symbol: KSR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173598.4 | NP_775869.3 | 4 | 862 | ||
| XM_011538224.2 | XP_011536526.1 | 4 | 1197 | |||
| XM_011538225.2 | XP_011536527.1 | 4 | 1656 | |||
| XM_011538226.2 | XP_011536528.1 | 4 | 1197 | |||
| XM_011538229.2 | XP_011536531.1 | 4 | 1197 | |||
| XM_017019208.1 | XP_016874697.1 | 4 | 1197 | |||
| XM_017019209.1 | XP_016874698.1 | 4 | 1197 | |||
| GenBank mRNA | AY345972.1 | - | 2 | 531 | ||
| BC107106.1 | - | 2 | 526 | |||
| BC107107.1 | - | 2 | 526 | |||
| BC127603.1 | - | 2 | 531 | 40019272 | ||

KSR2
Genomic Map