siRNA Details
| Gene Symbol: KSR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173598.4 | NP_775869.3 | 15 | 2265 | ||
| XM_011538224.2 | XP_011536526.1 | 15 | 2594 | |||
| XM_011538225.2 | XP_011536527.1 | 15 | 3059 | |||
| XM_011538228.2 | XP_011536530.1 | 13 | 1424 | |||
| XM_017019208.1 | XP_016874697.1 | 15 | 2600 | |||
| XM_017019210.1 | XP_016874699.1 | 10 | 1050 | |||
| GenBank mRNA | AY345972.1 | - | 13 | 1934 | ||
| BC107106.1 | - | 13 | 1929 | |||
| BC107107.1 | - | 13 | 1929 | |||
| BC127603.1 | - | 13 | 1934 | 40019272 | ||

KSR2
Genomic Map