siRNA Details
| Gene Symbol: GXYLT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099650.1 | NP_001093120.1 | 4 | 957 | ||
| NM_173601.1 | NP_775872.1 | 5 | 1050 | |||
| XM_017019211.1 | XP_016874700.1 | 5 | 789 | |||
| GenBank mRNA | AK125141.1 | - | 4 | 810 | ||
| AL832209.1 | - | 1 | 187 | |||
| BC015597.2 | - | 1 | 112 | |||
| BC030023.1 | - | 5 | 1058 | 5266607 | ||
| BC039145.1 | - | 4 | 797 | 4508300 | ||

GXYLT1
Genomic Map