siRNA Details
Gene Symbol: CHAMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164144.1 | NP_001157616.1 | 3 | 2068 | ||
NM_001164145.1 | NP_001157617.1 | 3 | 2032 | |||
NM_032436.2 | NP_115812.1 | 3 | 2071 | |||
GenBank mRNA | AB058705.1 | - | 1 | 1815 | ||
AK074894.1 | - | 1 | 1535 | |||
AK096346.1 | - | 3 | 2048 | |||
BC004820.1 | - | 1 | 18 | |||
BC065237.1 | - | 3 | 2021 | 6162118 |
