siRNA Details
| Gene Symbol: CHAMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164144.1 | NP_001157616.1 | 3 | 374 | ||
| NM_001164145.1 | NP_001157617.1 | 3 | 338 | |||
| NM_032436.2 | NP_115812.1 | 3 | 377 | |||
| GenBank mRNA | AB058705.1 | - | 1 | 121 | ||
| AK096346.1 | - | 3 | 354 | |||
| BC065237.1 | - | 3 | 327 | 6162118 | ||
| CN365659.1 | - | 3 | 278 | |||
| DA289328.1 | - | 3 | 343 | |||
| DB229502.1 | - | 3 | 336 | |||

CHAMP1
Genomic Map