siRNA Details
| Gene Symbol: SLC46A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135919.1 | NP_001129391.1 | 3 | 778 | ||
| NM_181785.3 | NP_861450.1 | 3 | 778 | |||
| XM_005266361.2 | XP_005266418.1 | 3 | 456 | |||
| GenBank mRNA | AF315594.1 | - | 3 | 362 | ||
| AK093932.1 | - | 2 | 1206 | |||
| AK124604.1 | - | 1 | 1470 | |||
| BC036662.1 | - | 3 | 740 | 4827143 | ||
| BC060850.1 | - | 3 | 599 | 4820254 | ||
| BC068556.1 | - | 3 | 606 | 4820792 | ||
| BX537565.1 | - | 3 | 589 | |||

SLC46A3
Genomic Map