siRNA Details
| Gene Symbol: LINC01588 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126497.1 | 4 | 1481 | |||
| NR_126498.1 | 4 | 2108 | ||||
| GenBank mRNA | AK091232.1 | - | 4 | 2107 | ||
| BC144351.1 | - | 4 | 1480 | 9052871 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01588| Gene Symbol: LINC01588 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126497.1 | 4 | 1481 | |||
| NR_126498.1 | 4 | 2108 | ||||
| GenBank mRNA | AK091232.1 | - | 4 | 2107 | ||
| BC144351.1 | - | 4 | 1480 | 9052871 | ||