siRNA Details
| Gene Symbol: C14orf178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173978.1 | NP_001167449.1 | 2 | 450 | ||
| NM_174943.3 | NP_777603.1 | 3 | 464 | |||
| GenBank mRNA | AK098842.1 | - | 4 | 709 | ||
| BC107147.2 | - | 2 | 306 | |||
| BC113929.1 | - | 3 | 378 | |||
| BC113986.1 | - | 2 | 306 | |||

C14orf178
Genomic Map