siRNA Details
| Gene Symbol: F2RL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256566.1 | NP_001243495.1 | 2 | 498 | ||
| NM_004101.3 | NP_004092.1 | 2 | 649 | |||
| GenBank mRNA | AK298585.1 | - | 2 | 498 | ||
| AK312848.1 | - | 2 | 537 | |||
| BC093648.1 | - | 2 | 544 | 7939493 | ||
| BC093650.1 | - | 2 | 544 | 7939495 | ||
| BC095499.1 | - | 2 | 442 | |||
| BX537386.1 | - | 2 | 630 | |||
| U92971.1 | - | 2 | 589 | |||

F2RL2
Genomic Map