siRNA Details
| Gene Symbol: C14orf177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182560.2 | NP_872366.2 | 4 | 737 | ||
| GenBank mRNA | AK098639.1 | - | 4 | 737 | ||
| BC093661.1 | - | 2 | 374 | 7939506 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C14orf177| Gene Symbol: C14orf177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182560.2 | NP_872366.2 | 4 | 737 | ||
| GenBank mRNA | AK098639.1 | - | 4 | 737 | ||
| BC093661.1 | - | 2 | 374 | 7939506 | ||