siRNA Details
| Gene Symbol: F3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001178096.1 | NP_001171567.1 | 4 | 772 | ||
| NM_001993.4 | NP_001984.1 | 4 | 772 | |||
| GenBank mRNA | AF487337.1 | - | 4 | 552 | ||
| AF497569.1 | - | 4 | 499 | |||
| AF497570.1 | - | 4 | 499 | |||
| AF497571.1 | - | 2 | 272 | |||
| AK312638.1 | - | 4 | 668 | |||
| AY940730.1 | - | 4 | 552 | |||
| AY940731.1 | - | 4 | 552 | |||
| BC011029.1 | - | 4 | 719 | 4073183 | ||
| BT019808.1 | - | 4 | 551 | |||
| CR541792.1 | - | 4 | 551 | |||
| J02931.1 | - | 4 | 662 | |||
| M16553.1 | - | 4 | 626 | |||
| M27436.1 | - | 4 | 650 | |||

F3
Genomic Map