siRNA Details
| Gene Symbol: HMGN2P46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_022014.1 | 6 | 900 | |||
| GenBank mRNA | AK096745.1 | - | 6 | 896 | ||
| AY271964.1 | - | 6 | 886 | |||
| AY271965.1 | - | 4 | 655 | |||
| AY271966.1 | - | 4 | 661 | |||
| AY271967.1 | - | 4 | 640 | |||

HMGN2P46
Genomic Map