siRNA Details
| Gene Symbol: REC114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042367.1 | NP_001035826.1 | 3 | 289 | ||
| XM_017022083.1 | XP_016877572.1 | 2 | 81 | |||
| GenBank mRNA | BC055412.1 | - | 3 | 270 | ||
| DB465854.1 | - | 3 | 289 | |||

REC114
Genomic Map