siRNA Details
| Gene Symbol: LOC283710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243538.1 | NP_001230467.1 | 3 | 440 | ||
| XM_011521463.2 | XP_011519765.1 | 3 | 439 | |||
| XM_011521465.2 | XP_011519767.1 | 3 | 439 | |||
| XM_017022084.1 | XP_016877573.1 | 3 | 439 | |||
| GenBank mRNA | BF974240.1 | - | 3 | 438 | ||
| BQ055725.1 | - | 2 | 319 | |||

LOC283710
Genomic Map