siRNA Details
Gene Symbol: LOC283710 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243538.1 | NP_001230467.1 | 4 | 695 | ||
GenBank mRNA | BF974240.1 | - | 4 | 692 | ||
BQ055725.1 | - | 3 | 574 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC283710 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243538.1 | NP_001230467.1 | 4 | 695 | ||
GenBank mRNA | BF974240.1 | - | 4 | 692 | ||
BQ055725.1 | - | 3 | 574 |