siRNA Details
| Gene Symbol: SAXO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008226.1 | NP_001008227.1 | 2, 3 | 319 | ||
| GenBank mRNA | AK304339.1 | - | 3, 4 | 431 | ||
| AL833762.1 | - | 2, 3 | 327 | |||
| BC144647.1 | - | 2, 3 | 541 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SAXO2| Gene Symbol: SAXO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008226.1 | NP_001008227.1 | 2, 3 | 319 | ||
| GenBank mRNA | AK304339.1 | - | 3, 4 | 431 | ||
| AL833762.1 | - | 2, 3 | 327 | |||
| BC144647.1 | - | 2, 3 | 541 | |||