siRNA Details
Gene Symbol: FAM169B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182562.2 | NP_872368.2 | 4 | 363 | ||
GenBank mRNA | AK097062.1 | - | 4 | 363 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FAM169B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182562.2 | NP_872368.2 | 4 | 363 | ||
GenBank mRNA | AK097062.1 | - | 4 | 363 |