siRNA Details
| Gene Symbol: C16orf54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175900.3 | NP_787096.2 | 2 | 1078 | ||
| GenBank mRNA | AK093000.1 | - | 2 | 1065 | ||
| AY358833.1 | - | 2 | 1062 | |||
| BC104466.2 | - | 2 | 1047 | 40032034 | ||
| BC104467.2 | - | 2 | 1047 | 40032037 | ||

C16orf54
Genomic Map