siRNA Details
| Gene Symbol: TMEM114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146336.1 | NP_001139808.1 | 4 | 843 | ||
| NM_001290095.1 | NP_001277024.1 | 3 | 705 | |||
| NM_001290097.1 | NP_001277026.1 | 4 | 797 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM114| Gene Symbol: TMEM114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146336.1 | NP_001139808.1 | 4 | 843 | ||
| NM_001290095.1 | NP_001277024.1 | 3 | 705 | |||
| NM_001290097.1 | NP_001277026.1 | 4 | 797 | |||