siRNA Details
| Gene Symbol: FADS6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178128.5 | NP_835229.3 | 6 | 1213 | ||
| XM_005257224.4 | XP_005257281.2 | 6 | 1207 | |||
| XM_017024458.1 | XP_016879947.1 | 7 | 988 | |||
| GenBank mRNA | AK094411.1 | - | 6 | 1061 | ||
| AK293720.1 | - | 3 | 572 | |||
| AY203929.1 | - | 6 | 1022 | |||
| BC117230.1 | - | 6 | 1042 | 40125781 | ||

FADS6
Genomic Map