siRNA Details
| Gene Symbol: HID1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030630.2 | NP_085133.1 | 5 | 742 | ||
| XM_005257226.1 | XP_005257283.1 | 5 | 737 | |||
| GenBank mRNA | AK074401.1 | - | 5 | 671 | ||
| AK095565.1 | - | 3 | 1930 | |||
| AK304081.1 | - | 5 | 626 | |||
| AL137556.1 | - | 5 | 686 | |||
| BC035372.1 | - | 5 | 658 | 5174235 | ||

HID1
Genomic Map