siRNA Details
| Gene Symbol: UBALD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182565.3 | NP_872371.1 | 3 | 1518 | ||
| GenBank mRNA | BC035511.2 | - | 3 | 1342 | 5091979 | |
| BU167469.1 | - | 1 | 486 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

UBALD2| Gene Symbol: UBALD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182565.3 | NP_872371.1 | 3 | 1518 | ||
| GenBank mRNA | BC035511.2 | - | 3 | 1342 | 5091979 | |
| BU167469.1 | - | 1 | 486 | |||