siRNA Details
| Gene Symbol: HEXDC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173620.2 | NP_775891.2 | 10 | 1471 | ||
| XM_005256346.3 | XP_005256403.1 | 9 | 1059 | |||
| XM_006722280.3 | XP_006722343.1 | 9 | 1034 | |||
| XM_011523560.2 | XP_011521862.1 | 8 | 1002 | |||
| XM_011523561.2 | XP_011521863.1 | 9 | 1040 | |||
| GenBank mRNA | BC035561.1 | - | 10 | 1152 | 5211583 | |

HEXDC
Genomic Map