siRNA Details
| Gene Symbol: VMO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144939.1 | NP_001138411.1 | 1 | 148 | ||
| NM_001144940.1 | NP_001138412.1 | 1 | 148 | |||
| NM_001144941.1 | NP_001138413.1 | 1 | 148 | |||
| NM_182566.2 | NP_872372.1 | 1 | 148 | |||
| GenBank mRNA | AF521892.1 | - | 1 | 153 | ||
| AY359099.1 | - | 1 | 78 | |||
| BC104195.1 | - | 1 | 116 | 40031205 | ||

VMO1
Genomic Map