siRNA Details
| Gene Symbol: C17orf58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181655.3 | NP_858041.2 | 2 | 397 | ||
| NM_181656.4 | NP_858042.2 | 2 | 397 | |||
| GenBank mRNA | AK026583.1 | - | 2 | 335 | ||
| BC021702.1 | - | 1 | 16 | |||
| BC036760.1 | - | 2 | 272 | 5587712 | ||
| BC105124.1 | - | 2 | 216 | 8144127 | ||
| BC112073.1 | - | 2 | 228 | 8327541 | ||
| BX364842.2 | - | 2 | 396 | |||

C17orf58
Genomic Map