siRNA Details
| Gene Symbol: C17orf105 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136483.1 | NP_001129955.1 | 3 | 249 | ||
| XM_011524633.1 | XP_011522935.1 | 4 | 1488 | |||
| XM_011524634.2 | XP_011522936.1 | 3 | 1588 | |||
| XM_011524635.1 | XP_011522937.1 | 3 | 1590 | |||
| XM_011524636.1 | XP_011522938.1 | 3 | 1590 | |||
| GenBank mRNA | AK125817.1 | - | 3 | 224 | ||
| BC054872.1 | - | 3 | 253 | |||
| BC070324.1 | - | 3 | 249 | |||
| BC146971.1 | - | 3 | 248 | 9056899 | ||

C17orf105
Genomic Map