siRNA Details
| Gene Symbol: FAM171A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198475.2 | NP_940877.2 | 6 | 958 | ||
| XM_017024490.1 | XP_016879979.1 | 4 | 525 | |||
| GenBank mRNA | BC029298.1 | - | 6 | 974 | ||
| BC030200.1 | - | 6 | 958 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM171A2| Gene Symbol: FAM171A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198475.2 | NP_940877.2 | 6 | 958 | ||
| XM_017024490.1 | XP_016879979.1 | 4 | 525 | |||
| GenBank mRNA | BC029298.1 | - | 6 | 974 | ||
| BC030200.1 | - | 6 | 958 | |||