siRNA Details
| Gene Symbol: TTLL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130918.1 | NP_001124390.1 | 10 | 1341 | ||
| NM_173623.3 | NP_775894.2 | 3 | 429 | |||
| XM_017024491.1 | XP_016879980.1 | 10 | 1484 | |||
| XM_017024492.1 | XP_016879981.1 | 9 | 1597 | |||
| XM_017024493.1 | XP_016879982.1 | 11 | 1551 | |||
| XM_017024494.1 | XP_016879983.1 | 10 | 1486 | |||
| XM_017024495.1 | XP_016879984.1 | 9 | 2214 | |||
| XM_017024496.1 | XP_016879985.1 | 5 | 624 | |||
| XM_017024497.1 | XP_016879986.1 | 10 | 1906 | |||
| GenBank mRNA | AK093127.1 | - | 2 | 293 | ||
| AK097392.1 | - | 10 | 1253 | |||
| AK302952.1 | - | 10 | 1287 | |||
| AK310321.1 | - | 3 | 439 | |||
| AL834151.1 | - | 10 | 1907 | |||
| BC033563.1 | - | 3 | 430 | 4825815 | ||
| BC041368.2 | - | 10 | 2337 | |||

TTLL6
Genomic Map