siRNA Details
| Gene Symbol: PIGW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178517.3 | NP_848612.2 | 2 | 863 | ||
| XM_005257238.2 | XP_005257295.1 | 2 | 1107 | |||
| XM_011524646.2 | XP_011522948.1 | 2 | 1405 | |||
| GenBank mRNA | AB097818.1 | - | 1 | 808 | ||
| AK094752.1 | - | 2 | 863 | |||

PIGW
Genomic Map