siRNA Details
| Gene Symbol: SLC13A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143838.2 | NP_001137310.1 | 6 | 912 | ||
| NM_001284509.1 | NP_001271438.1 | 6 | 861 | |||
| NM_001284510.1 | NP_001271439.1 | 5 | 783 | |||
| NM_177550.4 | NP_808218.1 | 6 | 912 | |||
| XM_011523795.2 | XP_011522097.1 | 6 | 902 | |||
| GenBank mRNA | AJ489980.1 | - | 5 | 695 | ||
| AK127797.1 | - | 6 | 809 | |||
| AK131206.1 | - | 3 | 1956 | |||
| AK172785.1 | - | 6 | 857 | |||
| AK297612.1 | - | 5 | 728 | |||
| AY151833.1 | - | 6 | 836 | |||
| BC104795.1 | - | 6 | 868 | 8143798 | ||
| BC112151.1 | - | 6 | 868 | 8327619 | ||
| BC143689.1 | - | 6 | 868 | 9052203 | ||
| CR749584.1 | - | 1 | 26 | |||

SLC13A5
Genomic Map