siRNA Details
| Gene Symbol: F11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000128.3 | NP_000119.1 | 5, 6 | 815 | ||
| XM_005262821.3 | XP_005262878.1 | 5, 6 | 834 | |||
| XM_005262822.3 | XP_005262879.1 | 5, 6 | 834 | |||
| XM_006714137.2 | XP_006714200.1 | 5, 6 | 834 | |||
| XM_017007884.1 | XP_016863373.1 | 5, 6 | 834 | |||
| XM_017007885.1 | XP_016863374.1 | 5, 6 | 834 | |||
| XM_017007886.1 | XP_016863375.1 | 5, 6 | 834 | |||
| GenBank mRNA | AK313195.1 | - | 5, 6 | 586 | ||
| BC119014.1 | - | 5, 6 | 578 | 40085167 | ||
| BC122863.1 | - | 5, 6 | 578 | 40085169 | ||
| M13142.1 | - | 5, 6 | 525 | |||

F11
Genomic Map