siRNA Details
| Gene Symbol: LOC101928710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110848.1 | 2 | 337 | |||
| NR_110849.1 | 2 | 446 | ||||
| GenBank mRNA | BG613706.1 | - | 2 | 447 | ||
| BX362577.2 | - | 2 | 331 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928710| Gene Symbol: LOC101928710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110848.1 | 2 | 337 | |||
| NR_110849.1 | 2 | 446 | ||||
| GenBank mRNA | BG613706.1 | - | 2 | 447 | ||
| BX362577.2 | - | 2 | 331 | |||