siRNA Details
| Gene Symbol: METRNL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004431.2 | NP_001004431.1 | 4 | 758 | ||
| XM_017024523.1 | XP_016880012.1 | 4 | 1107 | |||
| GenBank mRNA | AK093748.1 | - | 3 | 1329 | ||
| BC050568.1 | - | 3 | 488 | |||
| BC082252.1 | - | 4 | 758 | 6301543 | ||
| BC118558.1 | - | 4 | 245 | |||
| BC118638.1 | - | 4 | 245 | |||
| DR760693.1 | - | 3 | 478 | |||

METRNL
Genomic Map