siRNA Details
| Gene Symbol: SIGLEC15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213602.2 | NP_998767.1 | 2 | 217 | ||
| GenBank mRNA | AK095432.1 | - | 2 | 57 | ||
| AK172835.1 | - | 2 | 217 | |||
| AK290400.1 | - | 2 | 71 | |||
| AK290402.1 | - | 2 | 81 | |||
| AK301189.1 | - | 2 | 109 | |||
| AY971516.1 | - | 2 | 206 | |||
| DC299128.1 | - | 2 | 285 | |||

SIGLEC15
Genomic Map