siRNA Details
| Gene Symbol: SIGLEC15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213602.2 | NP_998767.1 | 6 | 1424 | ||
| GenBank mRNA | AK025833.1 | - | 1 | 71 | ||
| AK092746.1 | - | 3 | 975 | |||
| AK095432.1 | - | 7 | 1128 | |||
| AK172835.1 | - | 6 | 1424 | |||
| AY971516.1 | - | 6 | 1413 | |||

SIGLEC15
Genomic Map