siRNA Details
| Gene Symbol: SMIM21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037331.3 | NP_001032408.1 | 1 | 196 | ||
| NM_001303482.1 | NP_001290411.1 | 1 | 196 | |||
| XM_011525928.1 | XP_011524230.1 | 3 | 6506 | |||
| GenBank mRNA | AK093452.1 | - | 1 | 196 | ||
| BC037871.2 | - | 1 | 175 | |||
| BC105945.1 | - | 1 | 127 | |||
| BC107486.1 | - | 1 | 127 | 40032395 | ||

SMIM21
Genomic Map