siRNA Details
| Gene Symbol: ZNF776 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173632.3 | NP_775903.3 | 3 | 461 | ||
| GenBank mRNA | AK127764.1 | - | 2 | 107 | ||
| AK299680.1 | - | 3 | 461 | |||
| BC136752.1 | - | 2 | 103 | 9020742 | ||
| BC136753.1 | - | 2 | 103 | 9020743 | ||
| BX957217.1 | - | 3 | 416 | |||
| CR749390.1 | - | 3 | 416 | |||
| CR936802.1 | - | 3 | 415 | |||

ZNF776
Genomic Map