siRNA Details
| Gene Symbol: ZSCAN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182572.3 | NP_872378.3 | 6 | 1056 | ||
| XM_006723149.2 | XP_006723212.1 | 5 | 2669 | |||
| GenBank mRNA | AK091098.1 | - | 6 | 1056 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZSCAN1| Gene Symbol: ZSCAN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182572.3 | NP_872378.3 | 6 | 1056 | ||
| XM_006723149.2 | XP_006723212.1 | 5 | 2669 | |||
| GenBank mRNA | AK091098.1 | - | 6 | 1056 | ||