siRNA Details
| Gene Symbol: C19orf54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198476.3 | NP_940878.3 | 6 | 1224 | ||
| XM_005258776.4 | XP_005258833.1 | 6 | 1794 | |||
| XM_005258778.4 | XP_005258835.1 | 6 | 1213 | |||
| XM_017026626.1 | XP_016882115.1 | 7 | 1127 | |||
| XM_017026627.1 | XP_016882116.1 | 5 | 1680 | |||
| GenBank mRNA | AK310021.1 | - | 2 | 389 | ||
| BC090894.1 | - | 6 | 992 | 30377779 | ||

C19orf54
Genomic Map