siRNA Details
| Gene Symbol: CXCL17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198477.2 | NP_940879.1 | 2, 3 | 378 | ||
| GenBank mRNA | AK292985.1 | - | 2, 3 | 276 | ||
| AY358433.1 | - | 2, 3 | 314 | |||
| AY598464.1 | - | 2, 3 | 378 | |||
| BC093946.1 | - | 2, 3 | 229 | 7939791 | ||
| BC112095.1 | - | 2, 3 | 231 | 8327563 | ||

CXCL17
Genomic Map