siRNA Details
| Gene Symbol: CXCL17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198477.2 | NP_940879.1 | 4 | 536 | ||
| NR_133910.1 | 4 | 553 | ||||
| GenBank mRNA | AK292985.1 | - | 4 | 434 | ||
| AY358433.1 | - | 4 | 472 | |||
| AY598464.1 | - | 4 | 536 | |||
| BC093946.1 | - | 4 | 387 | 7939791 | ||
| BC112095.1 | - | 4 | 389 | 8327563 | ||

CXCL17
Genomic Map