siRNA Details
| Gene Symbol: ZNF575 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174945.2 | NP_777605.1 | 4 | 1660 | ||
| GenBank mRNA | AK057129.1 | - | 5 | 2236 | ||
| BC043611.1 | - | 4 | 1493 | 6018699 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF575| Gene Symbol: ZNF575 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174945.2 | NP_777605.1 | 4 | 1660 | ||
| GenBank mRNA | AK057129.1 | - | 5 | 2236 | ||
| BC043611.1 | - | 4 | 1493 | 6018699 | ||