siRNA Details
| Gene Symbol: F13B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001994.2 | NP_001985.2 | 11 | 1846 | ||
| XM_011509283.2 | XP_011507585.1 | 11 | 1843 | |||
| XM_011509284.2 | XP_011507586.1 | 11 | 1840 | |||
| XM_011509286.2 | XP_011507588.1 | 11 | 1854 | |||
| GenBank mRNA | AK290560.1 | - | 11 | 1843 | ||

F13B
Genomic Map