siRNA Details
| Gene Symbol: EMC10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175063.5 | NP_778233.4 | 8 | 1809 | ||
| NM_206538.3 | NP_996261.1 | 7 | 1722 | |||
| GenBank mRNA | AL512761.1 | - | 4 | 1325 | ||
| AY194293.2 | - | 12 | 2767 | |||
| AY358710.1 | - | 8 | 859 | |||
| AY761095.1 | - | 8 | 1787 | |||
| AY761097.1 | - | 7 | 1700 | |||
| BC032948.2 | - | 7 | 1661 | 4794992 | ||
| BC035001.2 | - | 7 | 1659 | 4796476 | ||
| BC062607.1 | - | 8 | 1951 | |||

EMC10
Genomic Map